Learn More
Medchemexpress LLC Picolinamide (2-pyridinecarboxamide) | 1452-77-3 | MFCD00023483 | 100.0% | 122.13 g/mol | C6H6N2O | 1 ML

Supplier: Medchemexpress LLC HY10102010MM 1ML
Picolinamide (2-picolinamide) is a small-molecule inhibitor of poly(ADP-ribose) synthetase (PARP) used as a research reagent for biochemical and cell-based studies. It is provided with a high-purity specification and available as a solid or as a ready-to-use solution for direct experimental use.
- Inhibitor of poly(ADP-ribose) synthetase (PARP).
- High purity (99.97% by HPLC) for research applications.
- Molecular formula C6H6N2O and molecular weight 122.13 g/mol.
- Available as solid and as 10 mM solution in DMSO (1 mL) for convenience.
- Stable storage: solid at room temperature (3 years); solutions recommended at -80°C or -20°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.