Learn More
Ansh Labs PAPP-A(pico) ELISA has a total 2 hours incubation, range of 6, 0.1-10 ng/mL, sensitivity of 0.037 ng/mL, sample of 50 µL Serum. Reacts with Human.
SDP
Supplier:  Ansh Labs AL101
Encompass
PAPP-A(pico) ELISA has a total 2 hours incubation, calibrators and range of 6, 0.1-10 ng/mL, sensitivity of 0.037 ng/mL, sample of 50 µL Serum. Reacts with Human. Crossreactivity with related molecules is not significant. The antibody pair used in the picoPAPP-A ELISA measures dimeric recombinant PAPP-A and PAPP-A/proMBP complex in equimolar concentration and does not cross react to PAPP-A2, MMP-9, and proMBP.picoPAPP A is useful in detection at low concentrations for studies of non-pregnant physiological states,pregnancy, dysfunctions of placental formation, certain cancers, and cardiovascular disease.Ansh Labs is a USA FDA registered manufacturer of diagnostic test kits, antibodies, antigens, and proteins.
Catalog No. NC1855409