Learn More
Enzo Life Sciences Picropodophyllin (1mg). CAS: 477-47-4
SDP
Supplier:  Enzo Life Sciences BMLEI3720001
Encompass_Preferred
A potent and selective inhibitor of insulin-like growth factor 1 receptor (IGF-1R, IC50=6 nM). It efficiently blocks IGF-1R activity, reduces pAKT and pERK1/2, induces apoptosis in IGF-1R-positive tumor cells and causes complete tumor regression in xenografted and allografted mice. Picropodophyllin downregulates IGF-1R by interfering with the action of β-arrestin 1/MDM2. It causes tumor regression and attenuates invasiveness of uveal melanoma cells. It displays beneficial effects on tumor growth, angiogenesis, bone disease and survival in a mouse multiple myeloma model. It attenuates intimal hyperplasia after vascular injury. Inhibition of IGF-1R is non-competitive with ATP. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (10mg/ml, warm) or dimethyl formamide (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0757