Learn More
Selleck Chemical LLC Picropodophyllin (PPP) 5mg 477-47-4 AXL1717
SDP
Supplier:  Selleck Chemical LLC S76685MG
Encompass_Preferred
Picropodophyllin (PPP, AXL1717) is a IGF-1R inhibitor with IC50 of 1 nM. It displays selectivity for IGF-1R and does not coinhibit tyrosine phosphorylation the IR, or of a selected panel of receptors less related to IGF-IR(FGF-R, PDGF-R, OR EGF-R). Picropodophyllin (PPP) induces apoptosis with antineoplastic activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0835767