Learn More
Medchemexpress LLC Picroside I | 27409-30-9 | 492.47 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN040710MM/1ML
Encompass_Preferred
Picroside I is the primary component of Picrorhiza scrophulariiflora, a valuable medicinal herb known for its abundant hepatoprotective metabolites, Picroside-I and Picroside-II. It shows promise as an agent for asthma management, significantly reducing inflammation at higher doses. Furthermore, Picroside I downregulates pSTAT6 and GATA3 expressions and dose-dependently increases serum levels of IFN-γ.
  • Major ingredient of Picrorhiza scrophulariiflora.
  • Source of hepatoprotective metabolites.
  • Promising agent for the management of asthma.
  • Reduces inflammation significantly at higher doses.
  • Downregulates pSTAT6 and GATA3 expressions.
  • Dose-dependently increases serum levels of IFN-γ.
Catalog No. 50-003-90030