Learn More
Medchemexpress LLC HY-N0407 5mg Medchemexpress, Picroside I CAS:27409-30-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN04075MG
Encompass_Preferred
Medchemexpress, HY-N0407 5mg Picroside I CAS:27409-30-9 Picroside I is the major ingredient of Picrorhiza kurroa. Picrorhiza kurroa is a high value medicinal herb due to rich source of hepatoprotective metabolites, Picroside-I and Picroside-II. Picroside I is a promising agent for the management of asthma. Picroside I reduces the inflammation significantly at its higher dose. Picroside I also downregulates pSTAT6 and GATA3 expressions. Picroside I dose-dependently increases the serum levels of IFN-γ. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0267