Learn More
Medchemexpress LLC Picrotin | 21416-53-5 | MFCD00868514 | 99.9% | 310.30 g/mol | C15H18O7 | 25 MG

Supplier: Medchemexpress LLC HY10778225MG
Picrotin is a picrotoxane sesquiterpenoid used in neuroscience research as an antagonist of GABAA and glycine receptors. Supplied as a solid research reagent, it is used in biochemical and pharmacological studies to probe inhibitory neurotransmission and receptor function.
- High purity (99.9%).
- Acts as an antagonist of GABAA and glycine receptors.
- White to off-white solid appearance.
- Molecular formula C15H18O7; molecular weight 310.30 g/mol.
- CAS number 21416-53-5 for unambiguous identification.
- Intended for laboratory and analytical use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.