Learn More
RAE Systems Inc PID SENSOR 0.1-5000PPM 0.1PPM
SDP
Supplier:  RAE Systems Inc C030912002/DEL
Encompass_Preferred
PID sensor (0.1 - 5,000 ppm; 0.1 ppm res.; 10.6 eV lamp)
Catalog No. NC0947295