Learn More
Fine Science Tools PIERSE FIXATION FORCEPS
SDP
Supplier:  Fine Science Tools 1815513
Encompass
Tip Shape: Straight/ Tips: Fixation/ Tip Dimensions: 0.1 x 0.4mm/ Tip Width: / Alloy / Material: Stainless Steel/ Length: 13cm
Catalog No. NC9100364