Learn More
Medchemexpress LLC HY-15484 10mg , Pifithrin-α (hydrobromide) CAS:63208-82-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1548410MG
Encompass_Preferred
HY-15484 10mg Pifithrin-α (hydrobromide) CAS: 63208-82-2 Pifithrin-α hydrobromide is a p53 inhibitor which blocks its transcriptional activity and prevents cells from apoptosis. Pifithrin-α hydrobromide is also an aryl hydrocarbon receptor (AhR) agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0166