Learn More
Medchemexpress LLC Pifithrin-β hydrobromide | 511296-88-1 | 99.97% | 349.29 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY16702A
Encompass_Preferred
Pifithrin-β hydrobromide (PFT β hydrobromide) is a potent p53 inhibitor with an IC50 of 23 μM. It is considered a lead compound for the treatment of cancer and neurodegenerative diseases.
  • Potent p53 inhibitor
  • Considered a lead compound for cancer and neurodegenerative disease therapy
  • Pifithrin-α quickly converts to pifithrin-β in culture medium
  • Pretreatments can exert neuroprotective effects after 24 hours
Catalog No. 50-003-95004