Learn More
Medchemexpress LLC Pifithrin-α, p-nitro, cyclic | 60477-38-5 | 99.4% | 299.35 g/mol | C15H13N3O2S | 50MG
SDP
Supplier:  Medchemexpress LLC HY12307650MG
Encompass_Preferred
Pifithrin-α, p-Nitro, Cyclic is a cell-permeable, active-form p53 inhibitor used in research to inhibit p53-dependent transcriptional activity and protect cells from p53-mediated apoptosis. It shows increased potency and a longer half-life compared with pifithrin-α, and demonstrates neuroprotective activity in etoposide-exposed cortical neurons (ED50 = 30 nM).
  • Cell-permeable active-form p53 inhibitor.
  • Higher potency than the parent compound; neuroprotective at low nanomolar concentrations.
  • Acts via post-transcriptional inhibition of p53 without preventing S15 phosphorylation.
  • High chemical purity suitable for research applications.
  • Supplied in milligram-scale quantities for laboratory use.
Catalog No. 50-002-37420