Learn More
Medchemexpress LLC Pifusertib hydrochloride | 1402602-94-1 | 98.7% | 424.49 g/mol | C26H25ClN4O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY19934A10MG
Encompass_Preferred
Pifusertib hydrochloride is the hydrochloride salt of TAS-117, a potent, selective, allosteric pan-AKT inhibitor supplied for preclinical and research use. It is provided as a high-purity solid with documented biochemical potency and has been investigated clinically for oncology indications.
  • Allosteric pan-AKT inhibitor with low-nanomolar activity against Akt isoforms.
  • High purity suitable for biochemical and cellular assays.
  • Hydrochloride salt form for improved stability and handling.
  • Molecular weight 424.49 g/mol; molecular formula C26H25ClN4O2.
  • Supplied in milligram quantities for preclinical studies.
  • Used in research on cancer biology, apoptosis, and autophagy.
Catalog No. 50-002-96788