Learn More
American Research Products Inc Pig Alpha fetoprotein AFP EL
Supplier:  American Research Products Inc CSBEL001421PI
Encompass
Pig Alpha-fetoprotein (AFP) ELISA kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 0 78 ng/mL Sample serum plasma tissue homogenates cell culture supernates Method Sandwich
Catalog No. 50-003-28536