Learn More
American Research Products Inc Pig calprotectin CALP ELISA
Supplier:  American Research Products Inc CSBEQ013485PI
Encompass
Pig calprotectin (CALP) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-27398