Learn More
American Research Products Inc Pig Interleukin 15 IL 15 ELI
Supplier:  American Research Products Inc CSBE14320P
Encompass
Pig Interleukin 15 (IL-15) ELISA Kit from CUSABIO Detection Range 0 78 pg/mL-50 pg/mL Sensitivity 0 195 pg/mL Sample serum plasma cell culture supernates tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-29457