Learn More
American Research Products Inc Pig N terminal pro brain natri
Supplier:  American Research Products Inc CSBEQ027465PI
Encompass
Pig N-terminal pro-brain natriuretic peptide (NT-proBNP) ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27865