Learn More
American Research Products Inc Pig thrombin antithrombin comp
Supplier:  American Research Products Inc CSBE13995P
Encompass
Pig thrombin-antithrombin compleX TAT ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28481