Learn More
Selleck Chemical LLC PIK-108 E2823-25MG
SDP
Supplier:  Selleck Chemical LLC E282325MG
Encompass_Preferred
PIK-108 is a non-ATP competitive allosteric p110 /p110 selective inhibitor
Catalog No. 50-000-40692