Learn More
Medchemexpress LLC PIK-75 | 372196-67-3 | 99.9% | 452.28 g/mol | C16H14BrN5O4S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY107834100MG
Encompass_Preferred
PIK-75 is a research-grade small-molecule inhibitor that selectively targets phosphoinositide 3-kinase p110α and DNA-dependent protein kinase (DNA-PK). It is supplied as a high-purity solid for biochemical and cell-based research and is commonly packaged in 100 mg amounts.
  • Reversible inhibitor of DNA-PK and p110α for biochemical and cell-based assays.
  • Potent activity with low-nanomolar to submicromolar IC50s against target kinases.
  • High purity (approx. 99.9%) suitable for research applications.
  • Solid powder form simplifies storage and handling.
  • Certificate of analysis and safety data sheet available to support quality and safety documentation.
Catalog No. 50-002-42802