Learn More
Medchemexpress LLC Pik-75 hydrochloride | 372196-77-5 | MFCD12546130 | 99.7% | 488.74 g/mol | C16H15BrClN5O4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY132815MG
Encompass_Preferred
PIK-75 hydrochloride is a reversible DNA-PK and p110α-selective inhibitor used in biochemical and cell-based research to probe PI3K signaling and DNA repair pathways. It shows nanomolar potency against DNA-PK and p110α, with reduced activity against other PI3K isoforms, and is supplied as a high-purity hydrochloride salt for laboratory research use only.
  • Nanomolar potency against DNA-PK and p110α.
  • Selectivity over other PI3K isoforms and related kinases.
  • High purity for biochemical and cell-based assays.
  • Solid hydrochloride salt, white to off-white appearance.
  • Stable under recommended storage; suitable for research use only.
Catalog No. 50-002-62399