Learn More
TARGETMOL CHEMICALS INC PIK-75 HYDROCHLORIDE 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T228750MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PIK-75 hydrochloride (PIK-75 HCl) is a p110(alpha) inhibitor with IC50 of 5.8 nM (200-fold more potently than p110(beta)) isoform-specific mutants at Ser773 and also potently inhibits DNA-PK with IC50 of 2 nM in cell-free assays. purity: 98%
Catalog No. 50-257-7175