Learn More
Selleck Chemical LLC PIK-93 25mg 593960-11-3
SDP
Supplier:  Selleck Chemical LLC S148925MG
Encompass_Preferred
PIK-93 is the first potent, synthetic PI4K (PI4KIII?) inhibitor with IC50 of 19 nM; shown to inhibit PI3K? with IC50 of 39 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-876-1