Learn More
Selleck Chemical LLC PIK-III 5mg 1383716-40-2 VPS34-IN2
SDP
Supplier:  Selleck Chemical LLC S76835MG
Encompass_Preferred
PIK-III (VPS34-IN2), which is a selective inhibitor of ?VPS34 enzymatic activity, inhibits autophagy and de novo lipidation of LC3 and leads to the stabilization of autophagy substrates. The IC50 values for VPS34 and PI(3)K? are 0.018 ?M and 1.2 ?M respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5124