Learn More
Selleck Chemical LLC Pilaralisib
SDP
Supplier:  Selleck Chemical LLC S76455MG
Encompass_Preferred
Pilaralisib (XL147) is a selective and reversible class I PI3K inhibitor for PI3K / / with IC50 of 39 nM/36 nM/23 nM in cell-free assays less potent to PI3K Phase 1/2
Catalog No. 50-000-01353