Learn More
Medchemexpress LLC Pilaralisib | 934526-89-3 | 99.14% | 541.02 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1652610MM/1ML
Encompass_Preferred
Pilaralisib is a potent and highly selective class I PI3Ks inhibitor with IC50s of 39 nM for PI3Kα, 383 nM for PI3Kβ, 23 nM for PI3Kγ, and 36 nM for PI3Kδ. It is also known as XL-147 and SAR245408.
  • Potent and highly selective class I PI3Ks inhibitor.
  • Inhibits PI3Kα, PI3δ, and PI3Kγ with IC50s of 39, 36, and 23 nM respectively.
  • Less potent against PI3Kβ (IC50 of 383 nM).
  • ATP-competitive inhibitor with a Ki value of 42 nM for PI3Kα.
  • Weak inhibitory activity toward VPS34 (IC50 of approximately 7.0 μM) and DNA-PK (IC50 of 4.75 μM).
  • Does not inhibit mTOR activity (IC50 > 15 μM).
  • Inhibits EGF-induced PIP3 production in PC-3 and MCF7 cells with IC50s of 220 and 347 nM.
  • Causes dose-dependent decrease in phosphorylation of AKT, p70S6K, and S6 in tumors in vivo.
Catalog No. 50-003-92196