Learn More
Medchemexpress LLC Pilocarpine | 92-13-7 | 99.9% | 1 ML

Supplier: Medchemexpress LLC HYB0726A10MM/1ML
Pilocarpine is a selective M3-type muscarinic acetylcholine receptor (M3 muscarinic receptor) agonist, intended for research use only. Studies have shown its effects both in vitro and in vivo.
- In vitro: induces dose- and time-dependent proliferation retardation and morphological abnormalities in human corneal stromal (HCS) cells at concentrations above 0.625 g/L.
- In vitro: evokes concentration-dependent relaxation with an EC50 of 2.4 mM in isolated segments of rat tail artery.
- In vivo: significantly increases saliva secretion in exercised rats compared to control rats.
- In vivo: leads to significantly lower Na+ concentration in the saliva of exercised rats compared to control rats.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.