Learn More
Medchemexpress LLC Pilocarpine Hydrochloride | 54-71-7 | MFCD00012722 | 99.92% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB072610G
Encompass_Preferred
Pilocarpine Hydrochloride is a potent M3-type muscarinic acetylcholine receptor (M3 muscarinic receptor) agonist. It is provided as a hydrochloride salt, derived from a natural alkaloid, and is primarily used for research purposes.
  • Potent M3-type muscarinic acetylcholine receptor agonist
  • Research use only
  • Stable for shipping at room temperature for less than two weeks
  • Store tightly sealed at 4°C, away from moisture and direct sunlight
Catalog No. 50-004-05563