Learn More
Medchemexpress LLC Pilocarpine nitrate | 148-72-1 | 99.8% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB100650MG
Encompass_Preferred
Pilocarpine nitrate is a potent agonist of the M3-type muscarinic acetylcholine receptor (M3 muscarinic receptor). This chemical, with 99.8% purity, appears as a white to off-white solid. It is intended for research use only.
  • Functions as a potent M3-type muscarinic acetylcholine receptor agonist.
  • Presents as a white to off-white solid.
  • Suitable for research involving M3 muscarinic receptor activity.
  • Can be used to study proliferation retardation and morphological abnormalities in human corneal stromal cells.
  • Demonstrates partial muscarinic agonist activity in isolated rat tail artery.
  • Useful for investigating effects on saliva secretion in exercised animal models.
  • Supports research into changes in ALOXE3 protein and arachidonic acid levels in mouse hippocampus during status epilepticus.
Catalog No. 50-004-07542