Learn More
Medchemexpress LLC Piloty's acid | 599-71-3 | ≥98.0% | 5 G

Supplier: Medchemexpress LLC HY1209905G
Piloty's acid (N-hydroxybenzenesulfonamide) is a crystalline biochemical reagent used as a nitroxyl (HNO) donor in research. It decomposes under basic conditions to release HNO and is commonly used in biochemical assays and studies of HNO chemistry.
- Purity ≥98%
- Physical form: crystalline solid
- Molecular weight 173.2 g/mol; formula C6H7NO3S
- Available in small laboratory sizes, including 5 G
- Store at -20°C to preserve stability
- Releases nitroxyl (HNO) more effectively above pH 8.0
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.