Learn More
Cayman Chemical Piloty s Acid Fatty Acids and
SDP
Supplier:  Cayman Chemical 1000699510 G
Encompass_Preferred
A well-characterized HNO donor; decomposes to HNO and benzenesulfinate anion; rate of HNO release at pH 7 is very slow (t½ = 5,500 minutes); most effective above pH 8.0; half-life decreases with an increase in pH to 561, 90, and 33 minutes at pH 8.0, 9.0, and 10.0, respectively; inhibits yeast aldehyde dehydrogenase (IC50 = 48 µM)
Catalog No. 50-289-0858