Learn More
Medchemexpress LLC PILR-alpha Human 1 100ug | 100ug
SDP
Supplier:  Medchemexpress LLC HYP77142100UG
Encompass_Preferred
Paired immunoglobulin-like type 2 receptor alpha (PILRA) is a immunoreceptor tyrosine-based inhibitory motif (ITIM)-bearing member of the paired receptor which consist of highly related activating and inhibitory receptors and are widely involved in the regulation of the immune system PILRA is thought to act as a cellular signaling inhibitory receptor by recruiting cytoplasmic phosphatases via their SH2 domains that block signal transduction such as SH1 through dephosphorylation of signaling molecules PILR-alpha Protein Human (178a a HEK293 His) is the recombinant human-derived PILR-alpha protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-86346