Learn More
Cayman Chemical PIlsIcaInIdehydrochLRIde 100mg
SDP
Supplier:  Cayman Chemical 34242100 MG
Encompass_Preferred
A class Ic antiarrhythmic agent; shortens action potential duration in canine Purkinje fibers, as well as increases the ratio of the effective refractory period to the duration of action potential in guinea pig atrial muscle strips, at 10 UG/ml; restores sinus rhythm in a canine model of ventricular multifocal arrhythmias from 3-10 mg/kg; suppresses arrhythmias induced by ouabain or halothane and epinephrine in dogs
Catalog No. 50-294-9543