Learn More
Cayman Chemical PIm-1 InhIbItr 2 1mg
SDP
Supplier:  Cayman Chemical 178091 MG
Encompass_Preferred
A potent Pim-1 inhibitor (Ki = 91 nM) that targets the ATP-binding kinase hinge region
Catalog No. 50-297-9885