Learn More
Novus Biologicals iCE280/iCE3 pI Marker 9.77, 200ul
SDP
Supplier:  Novus Biologicals 102219
Encompass
markers for pI determination that will give you both consistent and accurate measurement of isoelectric point on your iCE system. They can be used for whole-column and single-point capillary isoelectric focusing (cIEF) detection to measure your sample protein's isoelectric point, and for quality control of sample analysis. You'll get 200 µL of pI marker stock solution in every vial.
Catalog No. NC9572424