Learn More
TARGETMOL CHEMICALS INC PIMASERTIB 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T613150MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pimasertib (AS703026) is an orally bioavailable small-molecule inhibitor of MEK1 and MEK2 (MEK1/2) with potential antineoplastic activity. purity: 98%
Catalog No. 50-257-9532