Learn More
Selleck Chemical LLC Pimavanserin 50mg 706782-28-7 ACP-103
SDP
Supplier:  Selleck Chemical LLC S818350MG
Encompass_Preferred
Pimavanserin (ACP-103) is a potent and selective serotonin 5-HT2A inverse agonist with pIC50 of 8.73, used to treat psychosis associated with Parkinson¿s disease. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5765