Learn More
Selleck Chemical LLC Pimecrolimus 10mg 137071-32-0 ASM 981
SDP
Supplier:  Selleck Chemical LLC S500410MG
Encompass_Preferred
Pimecrolimus (ASM 981) is an immunophilin ligand, which binds specifically to the cytosolic receptor, immunophilin macrophilin-12 (FKBP-12); a calcineurin inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-741-2