Learn More
Medchemexpress LLC Pimelic acid monoethyl ester | 33018-91-6 | MFCD00014383 | 99.1% | 188.22 g/mol | C9H16O4 | 10 G

Supplier: Medchemexpress LLC HYW01608710G
Monoethyl pimelate is a small-molecule PROTAC linker and alkyl/ether building block used in medicinal chemistry for the synthesis of targeted protein degraders. It is the monoethyl ester of pimelic acid (CAS 33018-91-6), with formula C9H16O4 and molar mass 188.22 g/mol.
- Used as a PROTAC linker in degrader synthesis
- Suitable for synthesis of BCL-XL PROTAC degraders
- Molecular formula C9H16O4 and molecular weight 188.22 g/mol
- High chemical purity (~99.1%)
- Available as a 10 g pack for laboratory use
- Acts as an alkyl/ether building block for esterification chemistry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.