Learn More
eMolecules​ 74150-27-9 | 4,5-Dihydro-6-[2-(4-methoxyphenyl)-1H-benzimidazol-6-yl]-5-methyl-3(2H)-pyridazinone | Combi-Blocks | MFCD00761648 | 334.379 | C19H18N4O2 | 98.000 | COc1ccc(cc1)-c1nc2cc(ccc2[nH]1)C1=NNC(=O)CC1C | 1g | 267163138
Supplier:  eMolecules​ 267163138
Encompass_Preferred
4,5-Dihydro-6-[2-(4-methoxyphenyl)-1H-benzimidazol-6-yl]-5-methyl-3(2H)-pyridazinone | Combi-Blocks | 74150-27-9 | MFCD00761648 | 334.379 | C19H18N4O2 | 98.000 | COc1ccc(cc1)-c1nc2cc(ccc2[nH]1)C1=NNC(=O)CC1C | 1g | 267163138
Catalog No. 50-159-5472