Learn More
TARGETMOL CHEMICALS INC PIMOBENDAN 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T170825MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pimobendan (UD-CG 115 BS) is a selective inhibitor of PDE3 (IC50 0.32 uM). purity: 99%
Catalog No. 50-258-1126