Learn More
Medchemexpress LLC Pimobendan-d3 | 74150-27-9 | 99.4% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0204S15MG
Encompass_Preferred
Pimobendan-d3 (UD-CG115-d3) is deuterium labeled Pimobendan (HY-B0204), a selective inhibitor of PDE3 with an IC50 of 0.32 μM. It is intended for research use only and not sold to patients. This compound can be used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer and internal standard
  • Suitable for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled for potential impact on pharmacokinetic and metabolic profiles
  • Intended for research use only
Catalog No. 50-004-05957