Learn More
Medchemexpress LLC HY-105129 10mg Medchemexpress, Pimonidazole (hydrochloride) CAS:70132-51-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10512910MG
Encompass_Preferred
Medchemexpress, HY-105129 10mg Pimonidazole (HCl) CAS:70132-51-3 Pimonidazole is a novel hypoxia marker for complementary study of tumor hypoxia and cell proliferation in tumor. Pimonidazole accumulates in hypoxic cells via covalent binding with macromolecules or by forming reductive metabolites after reduction of its nitro group, it can be used for qualitative and quantitative assessment of tumor hypoxia . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8250