Learn More
Grainger Pin Gage .0080 Plus Class ZZ
Supplier:  Grainger 140G87
Encompass_Preferred
The Grainger Approved Pin Gage is a precision measuring instrument used to check the size of holes, slots, grooves and other internal dimensions. It has a hardened and precision ground gage pin that is 0.140 inches in diameter with a tolerance of +/- 0.0002 inches. The pin is made of 52100 alloy steel, heat treated to RC 60-62 hardness. It has a knurled grip handle and includes a plastic case for storage and protection. The pin gage allows for highly accurate measurements and quality control checks.
Catalog No. 50-274-9035