Learn More
Grainger Pin Gage .0480 Plus Class ZZ
Supplier:  Grainger 140H27
Encompass_Preferred
The Class X Go Plus Pin Gauges from Grainger are precision measuring tools used to check hole sizes. They are made from hardened tool steel and lapped to a tolerance of +/- .0001". The pin gauges have a knurled grip handle and come in sizes ranging from .011" to 1.000" in .001" increments. They can be used to accurately measure hole diameters in metal, plastic and other materials. The gauges are manufactured in the USA and each pin is serialized for traceability. They are available individually or in sets.
Catalog No. 50-274-9189