Learn More
Grainger Pin Gage .1070 Plus Class ZZ
Supplier:  Grainger 140H85
Encompass_Preferred
The Vermont Gage Pin Gage 140H85 is a precision pin gage with a diameter of 0.1400 inches. It is made from hardened and ground alloy steel for durability and accuracy. This pin gage has a knurled grip and carbide measuring surfaces for smooth handling and precise measurements. It can be used to accurately measure the size of holes, slots, and other precision openings. The gage has a tolerance of +/- 0.00004 inches to ensure reliable and repeatable measurements. This high quality pin gage is suitable for workshop, inspection, and production use.
Catalog No. 50-275-0747