Learn More
Grainger Pin Gage .7120 Minus Class ZZ
Supplier:  Grainger 140C95
Encompass_Preferred
The Grainger Approved Pin Gage is a precision tool for measuring holes, slots, and grooves. It features a 0.2500" diameter gage pin with a tolerance of +/- 0.0002", made from 52100 alloy steel, heat-treated to RC 60-62 hardness. The aluminum knurled grip ensures easy handling. Ideal for industrial applications, it serves as a reference for setting micrometers, calipers, and bore gauges. It includes a plastic storage case for protection. This high-quality pin gage ensures precise linear measurement.
Catalog No. 50-275-3426