Learn More
TARGETMOL CHEMICALS INC PIMONIDAZOLE HYDROCHLORI 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1653610MG
Encompass_Preferred
Also available in 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Pimonidazole hydrochloride accumulates in hypoxic cells via covalent binding with macromolecules or by forming reductive metabolites after the reduction of its nitro group. It can be used for qualitative and quantitative assessment of tumor hypoxia. Pimonidazole hydrochloride is a novel hypoxia marker for the complementary study of tumor hypoxia and cell proliferation in tumors. purity: 100%
Catalog No. 50-258-1169