Learn More
Chemglass Life Sciences Clamp, Pinch, Stainless Steel, #65
SDP
Supplier:  Chemglass Life Sciences CG15009
Encompass_Preferred
Clamp, Pinch, Stainless Steel, #65. Made entirely of stainless steel. Part numbers CG-150-01 and CG-150-03 are supplied WITHOUT a screw type locking device. All other sizes have the screw type locking device.Note: The size #102 Stainless Steel pinch clamp is available on a special order basis.
Catalog No. CG15009