Learn More
Medchemexpress LLC HY-N0549 100mg Medchemexpress, (-​)​-​α-​Pinene CAS:7785-26-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0549100MG
Encompass_Preferred
Medchemexpress, HY-N0549 100mg (-​)​-​α-​Pinene CAS:7785-26-4 (-)-α-Pinene is a monoterpene and shows sleep enhancing property through a direct binding to GABAA-benzodiazepine (BZD) receptors by acting as a partial modulator at the BZD binding site. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0283